missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-2,3-dimethylbutane 98.0+%, TCI America™

Catalog No. p-7126579
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10675G 5 g
D106725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10675G Supplier TCI America Supplier No. D10675G

Chemical Identifiers

CAS 594-81-0
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00053168
InChI Key KLFWZEFFWWOMIF-UHFFFAOYSA-N
Synonym sym-Dibromotetramethylethane, Pinacol Dibromide
PubChem CID 521884
IUPAC Name 2,3-dibromo-2,3-dimethylbutane
SMILES CC(C)(C(C)(C)Br)Br

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromo-2,3-dimethylbutane
TSCA No
Recommended Storage Refrigerator