missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1-propanol 98.0+%, TCI America™

Numéro de catalogue. p-7126433
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D079225G 25 g
D0792100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D079225G Fournisseur TCI America Code fournisseur D079225G
Il en reste null

Identifiants chimiques

CAS 75-84-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004682
InChI Key KPSSIOMAKSHJJG-UHFFFAOYSA-N
Synonym neopentyl alcohol, 2,2-dimethyl-1-propanol, 2,2-dimethylpropanol, tert-butylcarbinol, 1-propanol, 2,2-dimethyl, tert-butyl carbinol, neopentanol, neopentylalcohol, neoamyl alcohol, 2,2-dimethylpropyl alcohol
PubChem CID 6404
IUPAC Name 2,2-dimethylpropan-1-ol
SMILES CC(C)(C)CO

Spécifications

Melting Point 54°C
Color White
Boiling Point 114°C
Quantity 25 g
UN Number 1325
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethyl-1-propanol
EINECSNumber (2)-0217
TSCA Yes