missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(3,4-dimethylphenyl)hexafluoropropane 97.0+%, TCI America™

Catalog No. B14025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14025G Supplier TCI America Supplier No. B14025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 65294-20-4
Melting Point 78°C
Color White
Molecular Formula C19H18F6
Quantity 5 g
MDL Number MFCD00042167
Synonym 4,4′-(Hexafluoroisopropylidene)di-o-xylene
InChI Key GLFKFHJEFMLTOB-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)C(C2=CC(=C(C=C2)C)C)(C(F)(F)F)C(F)(F)F)C
IUPAC Name 4-[2-(3,4-dimethylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-1,2-dimethylbenzene
Molecular Weight (g/mol) 360.343
PubChem CID 103356
Formula Weight 360.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(3,4-dimethylphenyl)hexafluoropropane
Show More Show Less