missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Coverall Arc Flash Kit with Balaclava and Voltage Gloves (10)

Catalog No. 17001925
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001925 5X-Large
17001890 Small
17001895 Medium
17001900 Large
17001905 X-Large
17001910 2X-Large
17001915 3X-Large
17001920 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001925 Supplier National Safety Apparel™ Supplier No. KIT2CV11B5X09

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The premium choice for superior comfort, improved safety and above average durability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70E, NFPA 2112, CSA Z462
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 12 cal/cm²
Size 5X-Large
Material Cotton, Nylon
Color Navy
Suspender None
Ankle Style Open
Seam Type Stitched
Closure Type Zipper
Show More Show Less