missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoronaphthalene, 98%

Catalog No. AAA1837822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1837822 100 g
AAA1837806 5 g
AAA1837814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1837822 Supplier Thermo Scientific Chemicals Supplier No. A1837822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 321-38-0 | C10H7F | 146.164 g/mol

1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 321-38-0
Molecular Formula C10H7F
Molecular Weight (g/mol) 146.164
MDL Number MFCD00003873
InChI Key CWLKTJOTWITYSI-UHFFFAOYSA-N
Synonym fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
PubChem CID 9450
IUPAC Name 1-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2F

Specifications

Melting Point -10°C
Density 1.331
Boiling Point 215°C to 217°C
Flash Point 65°C (149°F)
Refractive Index 1.593
Quantity 100 g
Beilstein 1906413
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 146.17
Percent Purity 98%
Chemical Name or Material 1-Fluoronaphthalene
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 206-287-0
RTECSNumber QJ7100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only