missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopropane, 99%

Catalog No. AAA10461AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10461AU 1000 mL
AAA10461AK 250 mL
AAA104610J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA10461AU Supplier Thermo Scientific Chemicals Supplier No. A10461AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-94-5 | C3H7Br | 122.993 g/mol

1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

Solubility
Miscible with ethanol, water and diethyl ether.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 106-94-5
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.993
MDL Number MFCD00000254
InChI Key CYNYIHKIEHGYOZ-UHFFFAOYSA-N
Synonym n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
PubChem CID 7840
ChEBI CHEBI:47105
IUPAC Name 1-bromopropane
SMILES CCCBr

Specifications

Melting Point -110°C
Density 1.353
Boiling Point 70°C to 71°C
Flash Point 21°C (70°F)
Refractive Index 1.434
Linear Formula CH3CH2CH2Br
Quantity 1000 mL
UN Number UN2344
Beilstein 505936
Sensitivity Light sensitive
Merck Index 14,7845
Solubility Information Miscible with ethanol,water and diethyl ether.
Formula Weight 123
Percent Purity 99%
Chemical Name or Material 1-Bromopropane
Show More Show Less
Hazard Category H225-H315-H319-H335-H336-H350-H360FD-H373
Hazard Statement GHS H Statement
H225-H360-H373-H315-H319-H335-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOPROPANES
EINECSNumber 203-445-0
RTECSNumber TX4110000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only