missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctane, 99%

Catalog No. AC166815000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166815000 500 mL Glass bottle
AC166810025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166815000 Supplier Thermo Scientific Chemicals Supplier No. 166815000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-83-1 | C8H17Br | 193.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-83-1
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000276
InChI Key VMKOFRJSULQZRM-UHFFFAOYSA-N
Synonym n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
PubChem CID 8140
IUPAC Name 1-bromooctane
SMILES CCCCCCCCBr

Specifications

Melting Point -55.0°C
Density 1.1100g/mL
Boiling Point 201.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4508 to 1.4528
Linear Formula CH3(CH2)7Br
Quantity 500 mL
Beilstein 01,16
Merck Index 15,6853
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 193.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromooctane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 203-912-9

RUO – Research Use Only