missing translation for 'onlineSavingsMsg'
Learn More

1-Bromohexane, 99%

Catalog No. AAA1516622
Change view
Click to view available options
Quantity:
10,000 g
100 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1516622 100 g
AAA1516636 500 g
AAA151660E 2500 g
AAA151660C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1516622 Supplier Thermo Scientific Chemicals Supplier No. A1516622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111-25-1 | C6H13Br | 165.07 g/mol

1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromohexane is utilized in the preparation of pharmaceuticals and organic chemicals. It is involved in Grignard reactions as an alkylating agent and in the synthesis of bromo-cyclic compounds.

Solubility
Immiscible with water.

Notes
Keep away from heat and sources of ignition.

Chemical Identifiers

CAS 111-25-1
Molecular Formula C6H13Br
Molecular Weight (g/mol) 165.07
MDL Number MFCD00000271
InChI Key MNDIARAMWBIKFW-UHFFFAOYSA-N
Synonym hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga
PubChem CID 8101
IUPAC Name 1-bromohexane
SMILES CCCCCCBr

Specifications

Melting Point -85°C
Density 1.176
Boiling Point 156°C
Flash Point 50°C (122°F)
Odor Pleasant
Refractive Index 1.4475
Quantity 100 g
UN Number UN1993
Beilstein 1731290
Solubility Information Immiscible with water.
Formula Weight 165.08
Percent Purity 99%
Chemical Name or Material 1-Bromohexane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315
Flammable liquid and vapour.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-850-2
RTECSNumber MO0925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only