missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoheptane 98.0+%, TCI America™

Catalog No. B059825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B059825G 25 g
B0598500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B059825G Supplier TCI America Supplier No. B059825G

Chemical Identifiers

CAS 629-04-9
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00000273
InChI Key LSXKDWGTSHCFPP-UHFFFAOYSA-N
Synonym heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane
PubChem CID 12369
IUPAC Name 1-bromoheptane
SMILES CCCCCCCBr

Specifications

Melting Point -58°C
Color Yellow
Boiling Point 180°C
Quantity 25 g
UN Number 1993
Formula Weight 179.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromoheptane
EINECSNumber (2)-0067
RTECSNumber MI8100000
TSCA Yes