missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoheptane, 99%

Catalog No. AC107032500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107032500 250 mL Glass bottle
AC107030010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107032500 Supplier Thermo Scientific Chemicals Supplier No. 107032500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-04-9 | C7H15Br | 179.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-04-9
Molecular Formula C7H15Br
Molecular Weight (g/mol) 179.10
MDL Number MFCD00000273
InChI Key LSXKDWGTSHCFPP-UHFFFAOYSA-N
Synonym heptyl bromide, n-heptyl bromide, heptane, 1-bromo, 1-bromo heptane, bromoheptane, 1-bromo-heptane, labotest-bb ltbb001160, heptylbromide, 1bromoheptane, bromo-heptane
PubChem CID 12369
IUPAC Name 1-bromoheptane
SMILES CCCCCCCBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -58°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 180°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4489 to 1.4509
Linear Formula CH3(CH2)6Br
Quantity 250 mL
Beilstein 01, 155
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble
Formula Weight 179.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromoheptane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-068-8
RTECSNumber MI8100000
TSCA TSCA

RUO – Research Use Only