missing translation for 'onlineSavingsMsg'
Learn More

1-Bromododécane 98,0+%, TCI America™

Numéro de catalogue. B058725ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B058725ML 25 mL
B0587500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B058725ML Fournisseur TCI America Code fournisseur B058725ML

Identifiants chimiques

CAS 143-15-7
Molecular Formula C12H25Br
Molecular Weight (g/mol) 249.236
MDL Number MFCD00000225
InChI Key PBLNBZIONSLZBU-UHFFFAOYSA-N
Synonym dodecyl bromide, lauryl bromide, n-dodecyl bromide, dodecane, 1-bromo, bromododecane, 1-bromo dodecane, 1-bromo-dodecane, 1-bromo-n-dodecane, dodecane, bromo, unii-90t93tx09d
PubChem CID 8919
IUPAC Name 1-bromododécane
SMILES CCCCCCCCCCCCBr

Spécifications

Color Jaune
Boiling Point 140°C
Quantity 25 mL
Formula Weight 249.24
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material 1-Bromododecane
EINECSNumber (2)-0067
TSCA Yes