missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoadamantane 99.0+%, TCI America™

Catalog No. B039025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B039025G 25 g
B0390250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B039025G Supplier TCI America Supplier No. B039025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 768-90-1
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074721
InChI Key VQHPRVYDKRESCL-UHFFFAOYSA-N
Synonym 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane
PubChem CID 79106
IUPAC Name 1-bromoadamantane
SMILES C1C2CC3CC1CC(C2)(C3)Br

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromoadamantane