missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-methylcyclohexane, cis + trans, 97%

Catalog No. AAL0264006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0264006 5 g
AAL0264014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0264006 Supplier Thermo Scientific Chemicals Supplier No. L0264006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6294-40-2 | C7H13Br | 177.085 g/mol

1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-methylcyclohexane is used as intermediates in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6294-40-2
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.085
MDL Number MFCD00014282
InChI Key RLOHLPHAOHGRNM-UHFFFAOYSA-N
Synonym cyclohexane, 1-bromo-4-methyl, p-methylcyclohexyl bromide, 4-bromo-1-methylcyclohexane, 4-methylcyclohexyl bromide, acmc-1b9h5, 1-bromo-4-methyl-cyclohexane, 4-methylcyclohexyl bromide cis+trans, cyclohexane, 1-bromo-4-methyl-, trans, 1-bromo-4-methylcyclohexane, cis + trans
PubChem CID 22727
IUPAC Name 1-bromo-4-methylcyclohexane
SMILES CC1CCC(CC1)Br

Specifications

Density 1.262
Boiling Point 94°C to 96°C (50 mmHg)
Refractive Index 1.488
Quantity 5 g
Beilstein 1900954
Solubility Information Not miscible in water.
Formula Weight 177.09
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-methylcyclohexane, Cis + Trans
EINECSNumber 228-559-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only