missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-10,12-pentacosadiyne 96.0+%, TCI America™

Catalog No. B1340100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1340100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1340100MG Supplier TCI America Supplier No. B1340100MG
Only null left

Chemical Identifiers

CAS 94598-32-0
Molecular Formula C25H43Br
Molecular Weight (g/mol) 423.52
MDL Number MFCD00059029
InChI Key NUDHYRYEPCJLCL-UHFFFAOYSA-N
Synonym 10,12-Pentacosadiynyl Bromide
PubChem CID 15047027
IUPAC Name 1-bromopentacosa-10,12-diyne
SMILES CCCCCCCCCCCCC#CC#CCCCCCCCCCBr

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 423.52
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-10,12-pentacosadiyne
TSCA No
Recommended Storage Freezer