missing translation for 'onlineSavingsMsg'
Learn More

1,9-Dibromononane 97.0+%, TCI America™

Catalog No. D174625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D174625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D174625G Supplier TCI America Supplier No. D174625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4549-33-1
Molecular Formula C9H18Br2
Molecular Weight (g/mol) 286.051
MDL Number MFCD00000279
InChI Key WGAXVZXBFBHLMC-UHFFFAOYSA-N
Synonym nonane, 1,9-dibromo, nonamethylene dibromide, nonamethylene bromide, 1,9-dibromnonane, nonane,9-dibromo, 1,9-dibromononane, pubchem3900, acmc-1ai7y, ksc236g2b
PubChem CID 20677
IUPAC Name 1,9-dibromononane
SMILES C(CCCCBr)CCCCBr

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 288°C
Quantity 25 g
Formula Weight 286.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,9-Dibromononane