missing translation for 'onlineSavingsMsg'
Learn More

1,8-Dichlorohexadecafluorooctane 98.0+%, TCI America™

Catalog No. D28045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28045G 5 g
D280425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28045G Supplier TCI America Supplier No. D28045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 647-25-6
Molecular Formula C8Cl2F16
Molecular Weight (g/mol) 470.96
MDL Number MFCD00155747
InChI Key GFQXWORJCNTDPU-UHFFFAOYSA-N
Synonym 1,8-Dichloroperfluorooctane
PubChem CID 2774033
ChEBI CHEBI:38872
IUPAC Name 1,8-dichloro-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecafluorooctane
SMILES FC(F)(Cl)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Cl

Specifications

Color Colorless
Quantity 5 g
Formula Weight 470.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,8-Dichlorohexadecafluorooctane