missing translation for 'onlineSavingsMsg'
Learn More

1,7-Dibromoheptane 98.0+%, TCI America™

Catalog No. D211925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D211925G 25 g
D211910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D211925G Supplier TCI America Supplier No. D211925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4549-31-9
Molecular Formula C7H14Br2
Molecular Weight (g/mol) 258.00
MDL Number MFCD00000274
InChI Key LVWSZGCVEZRFBT-UHFFFAOYSA-N
Synonym heptane, 1,7-dibromo, heptamethylene dibromide, heptane,7-dibromo, 1,7-dibromo-heptane, pubchem3887, dibromo-1,7 heptane, acmc-209k2q, 1,7-dibromoheptane, ksc236m7h, lvwszgcvezrfbt-uhfffaoysa
PubChem CID 78309
IUPAC Name 1,7-dibromoheptane
SMILES BrCCCCCCCBr

Specifications

Color Yellow
Boiling Point 263°C
Quantity 25 g
Formula Weight 258.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,7-Dibromoheptane