missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromopropane 98.0+%, TCI America™

Catalog No. D0202500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0202500G 500 g
D020225G 25 g
D0202100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0202500G Supplier TCI America Supplier No. D0202500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-64-8
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.89
MDL Number MFCD00000255
InChI Key VEFLKXRACNJHOV-UHFFFAOYSA-N
Synonym trimethylene bromide, trimethylene dibromide, propane, 1,3-dibromo, alpha,gamma-dibromopropane, unii-yqr3048ix9, ccris 6711, .alpha.,.gamma.-dibromopropane, 1,3-dibromo propane, 1,3-dibromo-propane, .omega.,.omega.'-dibromopropane
PubChem CID 8001
IUPAC Name 1,3-dibromopropane
SMILES BrCCCBr

Specifications

Color Yellow
Boiling Point 167°C
Quantity 500 g
UN Number 3082
Formula Weight 201.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromopropane
EINECSNumber (2)-0059
RTECSNumber TX8575000
TSCA Yes