missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromobutane 98.0+%, TCI America™

Catalog No. D017525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D017525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D017525G Supplier TCI America Supplier No. D017525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-80-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000152
InChI Key XZNGUVQDFJHPLU-UHFFFAOYSA-N
Synonym butane, 1,3-dibromo, 1,3-butylene bromide, 1,3 dibromobutane, 1,3-dibrombutan, butane,1,3-dibromo, 1,3-dibromo-butan, 1,3-butylenebromide, 1,3-dibromo butane, 1,3-dibromo-butane, pubchem3866
PubChem CID 7889
IUPAC Name 1,3-dibromobutane
SMILES CC(CCBr)Br

Specifications

Color Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 215.92
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromobutane