missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromopropane 98.0+%, TCI America™

Catalog No. D0203500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0203500G 500 g
D020325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0203500G Supplier TCI America Supplier No. D0203500G
Only null left

Chemical Identifiers

CAS 78-75-1
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.889
MDL Number MFCD00000148
InChI Key XFNJYAKDBJUJAJ-UHFFFAOYSA-N
Synonym propylene dibromide, propane, 1,2-dibromo, propylene bromide, 2,3-dibromopropane, ccris 6710, ch2brchbrch3, propane, 1,2-dibromo-, r, propane, 1,2-dibromo-, s, 1,2 dibromopropane, 1,2-dibromo-propane
PubChem CID 6553
IUPAC Name 1,2-dibromopropane
SMILES CC(CBr)Br

Specifications

Melting Point -55°C
Color Yellow
Boiling Point 141°C
Quantity 500 g
Formula Weight 201.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dibromopropane
RTECSNumber TX8574000
TSCA Yes