missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromobutane, 98%

Catalog No. AAB2035630
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2035630 250 g
AAB2035618 50 g
AAB203560B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2035630 Supplier Thermo Scientific Chemicals Supplier No. B2035630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 533-98-2 | C4H8Br2 | 215.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 533-98-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000157
InChI Key CZWSZZHGSNZRMW-UHFFFAOYNA-N
Synonym butane, 1,2-dibromo, .alpha.-butylene dibromide, alpha-butylene dibromide, 1,2-dibomobutane, 1,2 dibromobutane, butane,2-dibromo, 1,2-dibromo-butane, alpha-butylene bromide, 1,2-butylene bromide, .alpha.-butylene bromide
PubChem CID 10792
IUPAC Name 1,2-dibromobutane
SMILES CCC(Br)CBr

Specifications

Melting Point -65°C
Density 1.789
Boiling Point 166°C to 167°C
Flash Point >110°C (230°F)
Refractive Index 1.5141
Quantity 250 g
Beilstein 1731380
Merck Index 14,1566
Formula Weight 215.93
Percent Purity 98%
Chemical Name or Material 1,2-Dibromobutane
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 208-581-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only