missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)pyrrole 97.0+%, TCI America™

Catalog No. B16225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16225G 5 g
B16221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16225G Supplier TCI America Supplier No. B16225G
Only null left

Chemical Identifiers

CAS 78358-86-8
Molecular Formula C6H8BrN
Molecular Weight (g/mol) 174.041
MDL Number MFCD00191322
InChI Key QBAVHEZVBGASER-UHFFFAOYSA-N
PubChem CID 11651225
IUPAC Name 1-(2-bromoethyl)pyrrole
SMILES C1=CN(C=C1)CCBr

Specifications

Color Yellow
Quantity 5 g
Formula Weight 174.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)pyrrole
TSCA No
Recommended Storage Refrigerator