missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-2-ethoxybenzene 96.0+%, TCI America™

Catalog No. B31915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31915G 5 g
B319125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31915G Supplier TCI America Supplier No. B31915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3-8-3259
Molecular Formula C10H13BrO2
Molecular Weight (g/mol) 245.12
MDL Number MFCD02030483
InChI Key IOYHGBZPUZBUTJ-UHFFFAOYSA-N
Synonym 2-2-ethoxyphenoxy ethyl bromide, 1-2-bromoethoxy-2-ethoxybenzene, 2-2-ethoxyphenoxy ethylbromide, 2-2-bromoethoxy phenetole, benzene, 1-2-bromoethoxy-2-ethoxy, 1-2-bromoethoxy-2-ethyoxybenzene, 1-2-bromoethoxy-2-ethoxy-benzene, 2-bromo-1-2-ethoxyphenoxy ethane, tamsulosin impurity i, pubchem13149
PubChem CID 7010183
IUPAC Name 1-(2-bromoethoxy)-2-ethoxybenzene
SMILES CCOC1=CC=CC=C1OCCBr

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
Formula Weight 245.12
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Bromoethoxy)-2-ethoxybenzene