missing translation for 'onlineSavingsMsg'
Learn More

1,10-Dibromodecane 95.0+%, TCI America™

Catalog No. D017925G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D017925G 25 g
D0179100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D017925G Supplier TCI America Supplier No. D017925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4101-68-2
Molecular Formula C10H20Br2
Molecular Weight (g/mol) 300.08
MDL Number MFCD00000222
InChI Key GTQHJCOHNAFHRE-UHFFFAOYSA-N
Synonym decamethylene dibromide, decane, 1,10-dibromo, 1,10-dibromodecan, 1,10-dibromo-decan, 1,10-decamethylene bromide, 1,10-decamethylene dibromide, a,w-dibromodecane, 1,10-dibromdecane, 1,10 dibromodecane, decane,10-dibromo
PubChem CID 221483
IUPAC Name 1,10-dibromodecane
SMILES BrCCCCCCCCCCBr

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 300.08
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,10-Dibromodecane
EINECSNumber (2)-0070
TSCA Yes