Get a Label Printer with Label Cartrdige Purchase
When you purchase 15 qualifying Bradyâ„¢ M4 or M5 label cartridges, you will receive a Brady M510 portable label printer at no additional cost.
View the qualifying and redeemable products below. After making your qualifying purchase, log into the Redemption Center to redeem this offer.
Features and Benefits
Brady M4 and M5 labels are specifically formulated for harsh laboratory conditions and adhere to irregular, rough, and curved surfaces.
Brady Chemical Resistant Cryogenic Polypropylene Labels
Read text and scan barcodes easily with high-performance B-7425 Chemical and Temperature Resistant Polypropylene that resists harsh lab processes.
Print Technology | Thermal Transfer |
---|---|
Printable Sides | Single-side Printable |
Description | Chemical Resistant Cryogenic Polypropylene Labels |
Length (Metric) | 7.6 m |
Color | Black on White |
Length (English) | 25 ft. |
Material | Polypropylene |
Sterility | Non-sterile |
Format | Cartridge (Label) |
Product Type | Label |
Temperature Range (English) | -112°F to +158°F |
Coating | None |
Temperature Range (Metric) | -80°C to +70°C |
Autoclavable | Autoclavable |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Brady Aggressive Adhesive Multi-Purpose Nylon Labels
Looking for a tough and versatile general-purpose label material? Look no further than the B-499 nylon cloth label material. Print almost anywhere with a Brady portable label printer.
Bradyâ„¢ QuickFlagâ„¢ Polyester Labels
Quickly flag wires and cables with the B-483 QuickFlagâ„¢ label material, featuring a tapered design to prevent mismatched edges. Print almost anywhere with a Bradyâ„¢ portable label printer.
Bradyâ„¢ FreezerBondzâ„¢ Ultra-Thin Cryogenic Polyester Labels
FreezerBondzâ„¢ Ultra-Thin Cryogenic Polyester Labels allow for labeling of curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material.
Print Technology | Thermal Transfer |
---|---|
Width (English) | 1 in |
Color | White |
Material | Polyester |
Width (Metric) | 25.40 mm |
Sterility | Non-sterile |
Thickness (Metric) | 0.081 mm |
Format | Die-cut |
Min. Operating Temperature (Metric) | -196°C |
For Use With (Equipment) | BMP51;BMP53;M511;M510 |
Product Type | Label |
Thickness (English) | 0.0032 in |
Temperature Range (English) | -320°F to 230°F |
Certifications/Compliance | 2011/65/EU, 2015/863 |
Temperature Range (Metric) | -196°C to 110°C |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ Aggressive Adhesive Multipurpose Nylon Vial and Tube Labels
The B-499 nylon cloth label is made of tough and versatile, general-purpose label material. Print almost anywhere with a Bradyâ„¢ portable label printer.
Format | Die-cut |
---|---|
Product Type | Label |
Temperature Range (English) | -40°F to +194°F |
Material | Nylon Cloth |
Sterility | Non-sterile |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ Color Polyester Laboratory Vial and Tube Labels
Easily differentiate lab samples with these B-494 color polyester labels, which are available in six color options. Pair with a versatile Bradyâ„¢ portable label printer and print almost anywhere.
Print Technology | Thermal Transfer |
---|---|
Width (English) | 1 in |
Quantity | 240 |
Material | Polyester |
For Use With (Application) | Cold temperature labelling;Laboratory labelling |
Width (Metric) | 25.40 mm |
Sterility | Non-sterile |
Thickness (Metric) | 0.0762 mm |
Format | Die-cut |
Min. Operating Temperature (Metric) | -196°C |
For Use With (Equipment) | BMP51;BMP53;M511;M510 |
Product Type | Label |
Diameter (Metric) | 9.53 mm |
Thickness (English) | 0.003 in |
Temperature Range (English) | -320°F to 158°F |
No. per Pack | 240 Sets / Box |
Certifications/Compliance | 2011/65/EU, 2015/863 |
Temperature Range (Metric) | -196°C to 70°C |
Autoclavable | Not Autoclavable |
Finish | Gloss |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ All-weather Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Bradyâ„¢ handheld label maker.
Print Technology | Thermal Transfer |
---|---|
Printable Sides | Top |
Quantity | 1 |
Material | Vinyl |
Height (English) Printable Area | 0.5 in. |
For Use With (Application) | Component and equipment labelling;Electrical labelling;Facility identification;Floor marking;Lean and 5S labelling;Pipe and valve marking;Warehouse marking |
Sterility | Non-sterile |
Thickness (Metric) | 0.102 mm |
Format | Continuous |
Min. Operating Temperature (Metric) | -40°C |
Product Type | Label |
Thickness (English) | 0.004 in |
Temperature Range (English) | -40°F to 180°F |
Coating | None |
Certifications/Compliance | BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863 |
Temperature Range (Metric) | -40°C to 82°C |
Autoclavable | Not Autoclavable |
Finish | Gloss |
Adhesive Type | Permanent Acrylic |
Brady All Weather Permanent Adhesive Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label maker.
Bradyâ„¢ Solvent Resistant Polyester Labels
Easily label lab slides and well plates with the B-488 high-performance polyester label material. Print almost anywhere with a durable Bradyâ„¢ portable label maker.
Print Technology | Thermal Transfer |
---|---|
Printable Sides | Top |
Format | Die-cut |
Product Type | Label |
Color | White |
Material | Polyester |
Coating | None |
Height (English) Printable Area | 0.5 in. |
Autoclavable | Autoclavable |
Sterility | Non-sterile |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ Aggressive Adhesive, Multipurpose Clear Polyester Labels
Label circuit boards, panels and equipment with the B-432 clear polyester label material, allowing the applied surface colors to transmit around the printed label text. Print almost anywhere with a Bradyâ„¢ handheld label maker.
Width (English) | 1 in |
---|---|
Color | Transparent,Clear |
Material | Polyester |
For Use With (Application) | Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;Electrical labelling;General identification;Inventory and inspection labelling;Warehouse marking |
Width (Metric) | 25.40 mm |
Sterility | Non-sterile |
Thickness (Metric) | 0.102 mm |
Format | Die-cut |
For Use With (Equipment) | BMP51;BMP53;M511;M510 |
Product Type | Label |
Thickness (English) | 0.004 in |
Temperature Range (English) | -40°F to 212°F |
Certifications/Compliance | UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863 |
Temperature Range (Metric) | -40°C to 100°C |
Finish | Gloss |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ BradyGripâ„¢ Print-on-Hook Labels featuring VELCROâ„¢ Brand Hook
Easily place, remove and reposition labels on cable bundles with the BradyGripâ„¢ Print-on Hook label material (B-414), paired with VELCROâ„¢ Brand ONE-WRAPâ„¢ Rolls. Available for Bradyâ„¢ portable label printers.
Length (Metric) | 3.66 m |
---|---|
Color | White |
Length (English) | 12 ft. |
Quantity | 1 |
Material | Polyester |
For Use With (Application) | Wire and cable labelling |
Sterility | Non-sterile |
Thickness (Metric) | 0.381 mm |
Format | Continuous |
For Use With (Equipment) | BMP41;BMP51;BMP53;M511;M410;M510 |
Product Type | Label |
Thickness (English) | 0.015 in |
Temperature Range (English) | -40°F to 122°F |
Certifications/Compliance | 2011/65/EU, 2015/863 |
Temperature Range (Metric) | -40°C to 50°C |
Finish | Matte |
Adhesive Type | Non-adhesive |
Bradyâ„¢ FreezerBondzâ„¢ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Bradyâ„¢ handheld label maker.
Print Technology | Thermal Transfer |
---|---|
Product Type | Label |
Color | White |
Temperature Range (English) | -320°F to 230°F |
Material | Polyester |
Sterility | Non-sterile |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ PermaSleeveâ„¢ Heat-shrink Labels
Permanently mark wires with the durable PermaSleeveâ„¢ PS Heat-shrink polyolefin sleeve label material (B-342). Designed for use with a durable Bradyâ„¢ handheld label printer.
Product Type | Wire and Cable Markers |
---|---|
Temperature Range (English) | -67°F to 275°F |
Material | Polyolefin |
Certifications/Compliance | UL Recognised to UL224;SAE-AMS-DTL-23053/5 C (Class 1);SAE AS-81531 Compliant with Directive 2011/65/EU and 2015/863 |
Temperature Range (Metric) | -55°C to 135°C |
Sterility | Non-sterile |
Finish | Matte |
Adhesive Type | Non-adhesive |
Bradyâ„¢ Self-laminating Vinyl Wrap Around Labels
Mark cables and wires with the B-427 self-laminating vinyl label material, designed with a clear self-laminating protective layer. Print almost anywhere with a durable Bradyâ„¢ portable label printer.
Thickness (Metric) | 0.102 mm |
---|---|
Product Type | Label |
Thickness (English) | 0.004 in |
Temperature Range (English) | -94°F to 158°F |
Material | Vinyl |
Certifications/Compliance | UL Recognised to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL Recognised B-427 labels. Brady sells both UL Recognised B-427 labels and non-approved labels depending on customer requirements. Compliant with Directive 2011/65/EU and 2015/863 |
Temperature Range (Metric) | -70°C to 70°C |
Sterility | Non-sterile |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Bradyâ„¢ Metalized Matte Gray Polyester Labels
Create tough serial and rating plate labels with the B-428 metalized polyester label material with flexible high-tack adhesive. Designed for use with durable Bradyâ„¢ portable label makers.
Thickness (Metric) | 0.122 mm |
---|---|
Format | Die-cut |
Product Type | Label |
Thickness (English) | 0.0047 in |
Color | Light Gray |
Temperature Range (English) | -40°F to 248°F |
Material | Polyester |
Certifications/Compliance | UL Recognised;CSA Accepted;AGA Approved;Halogen Free (DIN VDE 0472/part 815) Compliant with Directive 2011/65/EU and 2015/863 |
Temperature Range (Metric) | -40°C to 120°C |
For Use With (Application) | Asset tracking;Circuit board labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling |
Sterility | Non-sterile |
Finish | Matte |
Adhesive Type | Permanent Acrylic |
Catalog Number | Description | |||
---|---|---|---|---|
Catalog Number |
Description Brady M510 Portable Label Printer | Learn More |