Learn More
Brady™ BradyGrip™ Print-on-Hook Labels featuring VELCRO™ Brand Hook
Easily place, remove and reposition labels on cable bundles with the BradyGrip™ Print-on Hook label material (B-414), paired with VELCRO™ Brand ONE-WRAP™ Rolls. Available for Brady™ portable label printers.
$58.56 - $89.82
Specifications
Certifications/Compliance 2011/65/EU, 2015/863
Adhesive Type Non-adhesive
Color White
Finish Matte
Format Continuous
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (LxW) Price Quantity  
Catalog Number Mfr. No. Dimensions (LxW) Price Quantity  
17-012-198
View Documents Promotion Details
Brady™
M4C250414
6.35 mm (W) x 3.66 m (L)
Each for $58.56
Only null left
17-012-146
View Documents Promotion Details
Brady™
M4C500414
12.70 mm (W) x 3.66 m (L)
Each for $67.06
Only null left
17-012-297
View Documents Promotion Details
Brady™
M4C750414
19.05 mm (W) x 3.66 m (L)
Each for $89.82
Only null left
Description
Description
  • Designed in partnership with Velcro™ Companies to easily and safely mark wire and cable bundles
  • Constructed for long-term use and reuse
  • Made with Velcro™ Brand hooks for secure placement on VELCRO™ Brand ONE-WRAP™ Straps
  • Easy to reposition and reuse when you need to update your bundles
  • VELCRO™ Brand ONE-WRAP™ product not included with purchase
  • VELCRO™ and ONE-WRAP™ are registered trademarks of Velcro™ IP Holdings, LLC
Specifications
Specifications
2011/65/EU, 2015/863
White
Continuous
Label
Wire and cable labelling
1
-40°C to 50°C
0.381 mm
3.66 m
Non-adhesive
Matte
Polyester
Non-sterile
BMP41;BMP51;BMP53;M511;M410;M510
-40°F to 122°F
0.015 in
12 ft.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'