UserName
HealthPRO Membership Benefits
HealthPRO Membership Benefits
With our proven record of customer service and support, the Fisher Scientific channel is your trusted source for laboratory medicine.
Customer Service
If you do not have a Fisher Scientific account or have difficulty signing in to your business account, please contact Customer Service at 1‑800-234-7437 or visit the Help & Support Center.
HealthPRO Canada Logo
Featured Products
Mopec 5000 Autopsy Saw
Mopec 5000 Autopsy Saw
The Mopec 5000 Autopsy Saw is developed specifically for autopsy use and is hermetically sealed and CE certified. It’s totally submersible for complete cleaning and offers a cool touch, reduced vibration, and safe installation.
Fisherbrand Histoplast Paraffin Wax
Fisherbrand Histoplast Paraffin Wax
Fisherbrand™ Histoplast Paraffin Wax is made from high-purity raw materials and provides consistent tissue ribbons every time. Choose Histoplast PE for general use, Histoplast IM for use on large and dense specimens, and Histoplast LP (formulated for a lower melting point) for improved ribboning with less compression.
MICROMESH Biopsy Processing/Embedding Cassettes
MICROMESH Biopsy Processing/Embedding Cassettes
Simport™ Scientific MICROMESH™ Biopsy Processing/Embedding Cassettes feature a large, 27 mm square compartment, suitable even for needle biopsies. They maximize fluid exchange and drainage and can be opened and closed as often as necessary without danger of specimen loss.
Top Brands
The HealthPRO and Fisher Scientific Partnership
If you have any questions about the HealthPRO and Fisher Scientific partnership, reach out to us using the form on this page.
We’re your trusted source for:
  • Lab consumables, supplies, chemicals, and safety products
  • Cassettes and embedding
  • Microscope slides
Together, HealthPRO and the Fisher Scientific channel bring you:
  • An industry-leading portfolio with more than 1.7 million items, including 80,000 exclusive products and more than 9,400 Fisherbrand™ alternatives
  • Guaranteed savings
  • Attractive participation incentives
Contact Us
Fill out the form below to learn more about the HealthPRO and Fisher Scientific partnership.
Required Field *