CAS RN 998-41-4
CAS RN 998-41-4
IUPAC Name:
tributylsilyl
Synonyms:
tributylsilyl group
Molecular Weight (g/mol):
199.43
Molecular Formula:
C12H27Si
InChi Key:
ISEIIPDWJVGTQS-UHFFFAOYSA-N
SMILES:
CCCC[Si](CCCC)CCCC
1
–
1
of
1
results
1
Tributylsilane 98.0+%, TCI America™
CAS: 998-41-4 Molecular Formula: C12H27Si Molecular Weight (g/mol): 199.43 MDL Number: MFCD00009415 InChI Key: ISEIIPDWJVGTQS-UHFFFAOYSA-N PubChem CID: 6327711 IUPAC Name: tributylsilyl SMILES: CCCC[Si](CCCC)CCCC