accessibility menu, dialog, popup

UserName

CAS RN 99464-83-2

IUPAC Name: 1-chloroethyl cyclohexyl carbonate
Synonyms: 1-chloroethyl cyclohexyl carbonate
Molecular Weight (g/mol): 206.67
Molecular Formula: C9H15ClO3
InChi Key: ONZWFHWHTYZZLM-UHFFFAOYNA-N
SMILES: CC(Cl)OC(=O)OC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results