missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroethyl Cyclohexyl Carbonate 98.0+%, TCI America™

Catalog No. C178625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C178625G 25 g
C1786250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C178625G Supplier TCI America Supplier No. C178625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99464-83-2
Molecular Formula C9H15ClO3
Molecular Weight (g/mol) 206.666
MDL Number MFCD04038149
InChI Key ONZWFHWHTYZZLM-UHFFFAOYSA-N
Synonym Carbonic Acid 1-Chloroethyl Cyclohexyl Ester
PubChem CID 11229529
IUPAC Name 1-chloroethyl cyclohexyl carbonate
SMILES CC(OC(=O)OC1CCCCC1)Cl

Specifications

Color Yellow
Boiling Point 107°C
Quantity 25 g
UN Number 1760
Formula Weight 206.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl Cyclohexyl Carbonate