accessibility menu, dialog, popup

UserName

CAS RN 99365-40-9

IUPAC Name: 6-bromo-2,3-dihydro-1H-indol-2-one
Synonyms: 6-bromo-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 212.05
Molecular Formula: C8H6BrNO
InChi Key: JARRYVQFBQVOBE-UHFFFAOYSA-N
SMILES: BrC1=CC=C2CC(=O)NC2=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)