accessibility menu, dialog, popup

UserName

HClH3CONCH3H3CHO

CAS RN 99300-78-4

IUPAC Name: hydrogen 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol chloride
Synonyms: venlafaxine hcl venlafaxine hydrochloride
Molecular Weight (g/mol): 313.87
Molecular Formula: C17H28ClNO2
InChi Key: QYRYFNHXARDNFZ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC1=CC=C(C=C1)C(CN(C)C)C1(O)CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)