accessibility menu, dialog, popup

UserName

FOOOOH

CAS RN 99199-59-4

IUPAC Name: 6-fluoro-4-oxo-4H-chromene-2-carboxylic acid
Synonyms: 6-fluoro-4-oxochromene-2-carboxylic acid
Molecular Weight (g/mol): 208.14
Molecular Formula: C10H5FO4
InChi Key: JZJYDFADRMBXAW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)C2=CC(F)=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results