missing translation for 'onlineSavingsMsg'
Learn More

6-Fluorochromone-2-carboxylic Acid 97.0+%, TCI America™

Catalog No. F07445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07445G 5 g
F074425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07445G Supplier TCI America Supplier No. F07445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99199-59-4
Molecular Formula C10H5FO4
Molecular Weight (g/mol) 208.144
MDL Number MFCD03070543
InChI Key JZJYDFADRMBXAW-UHFFFAOYSA-N
Synonym 6-fluorochromone-2-carboxylic acid, 6-fluoro-4-oxo-4h-chromene-2-carboxylic acid, 6-fluoro-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo, 6-fluorochromone-2-carboxylicacid, pubchem9710, acmc-209sbz
PubChem CID 4096046
IUPAC Name 6-fluoro-4-oxochromene-2-carboxylic acid
SMILES C1=CC2=C(C=C1F)C(=O)C=C(O2)C(=O)O

Specifications

Melting Point 258°C
Color White-Yellow
Quantity 5 g
Formula Weight 208.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluorochromone-2-carboxylic Acid