accessibility menu, dialog, popup

UserName

H3CCH3CH3

CAS RN 99-86-5

IUPAC Name: 1-methyl-4-(propan-2-yl)cyclohexa-1,3-diene
Synonyms: terpinene terpilene α-terpinene α-terpinen
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: YHQGMYUVUMAZJR-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)CC1
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)