missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpinene 90.0+%, TCI America™

Catalog No. M0317500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0317500ML 500 mL
M031725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0317500ML Supplier TCI America Supplier No. M0317500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001534
InChI Key YHQGMYUVUMAZJR-UHFFFAOYSA-N
Synonym alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
PubChem CID 7462
ChEBI CHEBI:10334
IUPAC Name 1-methyl-4-(propan-2-yl)cyclohexa-1,3-diene
SMILES CC(C)C1=CC=C(C)CC1

Specifications

Color Yellow
Boiling Point 173°C
Quantity 500 mL
UN Number 2319
Formula Weight 136.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Terpinene
EINECSNumber (3)-2244
RTECSNumber OS8060000
TSCA Yes
Recommended Storage Refrigerator