accessibility menu, dialog, popup

UserName

CAS RN 99-73-0

IUPAC Name: 2-bromo-1-(4-bromophenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-bromophenyl)ethanone
Molecular Weight (g/mol): 277.94
Molecular Formula: C8H6Br2O
InChi Key: FKJSFKCZZIXQIP-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)