missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenacyl Bromide 99.0+%, TCI America™

Catalog No. A55015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A55015G Supplier TCI America Supplier No. A55015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-73-0
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00000200
InChI Key FKJSFKCZZIXQIP-UHFFFAOYSA-N
Synonym 2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
PubChem CID 7454
IUPAC Name 2-bromo-1-(4-bromophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)Br

Specifications

Melting Point 111°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 277.94
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenacyl Bromide [for HPLC Labeling]
EINECSNumber (3)-2935
RTECSNumber AM6950000
TSCA Yes
Recommended Storage Refrigerator