accessibility menu, dialog, popup

UserName

HOOONHHO

CAS RN 99-11-6

IUPAC Name: 6-hydroxy-2-oxo-1,2-dihydropyridine-4-carboxylic acid
Synonyms: citrazinic acid
Molecular Weight (g/mol): 155.11
Molecular Formula: C6H5NO4
InChi Key: CSGQJHQYWJLPKY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=O)NC(O)=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)