missing translation for 'onlineSavingsMsg'
Learn More

Citrazinic acid, 98%

Catalog No. AC167011000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167011000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC167011000 Supplier Thermo Scientific Chemicals Supplier No. 167011000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-11-6 | C6H5NO4 | 155.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-11-6
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00006274
InChI Key CSGQJHQYWJLPKY-UHFFFAOYSA-N
Synonym citrazinic acid, 2,6-dihydroxyisonicotinic acid, 2,6-dihydroxypyridine-4-carboxylic acid, 2,6-dihydroxy-4-carboxypyridine, 2,6-dihydroxy-4-pyridinecarboxylic acid, kyselina citrazinova, 2,6-dihydroxyisonicotinate, 4-pyridinecarboxylic acid, 2,6-dihydroxy, citrazinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-6-hydroxy-2-oxo
PubChem CID 7425
IUPAC Name 2-hydroxy-6-oxo-1H-pyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(=O)NC(O)=C1

Specifications

Melting Point >300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (HO)2(C5H2N)COOH
Quantity 100 g
Beilstein 22, 254
Merck Index 15, 2324
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alkali hydroxide or carbonate solns.,slightly soluble in hot hcl
Formula Weight 155.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Citrazinic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-731-2

RUO – Research Use Only