accessibility menu, dialog, popup

UserName

CAS RN 98619-07-9

H3COOHF

CAS RN 98619-07-9

IUPAC Name: 1-(2-fluoro-4-hydroxyphenyl)ethan-1-one
Synonyms: 1-(2-fluoro-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: ZCZIRBNZMFUCOH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)