missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-hydroxyacetophenone, 97%

Catalog No. AAA1988414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1988414 25 g
AAA1988406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1988414 Supplier Thermo Scientific Chemicals Supplier No. A1988414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98619-07-9 | C8H7FO2 | 154.14 g/mol

2'-Fluoro-4'-hydroxyacetophenone is used in used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-4′-hydroxyacetophenone is used in used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 98619-07-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00142713
InChI Key ZCZIRBNZMFUCOH-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-hydroxyacetophenone, 1-2-fluoro-4-hydroxyphenyl ethanone, 4-hydroxy-2-fluoroacetophenone, 2-fluoro-4-hydroxyacetophenone, 1-2-fluoro-4-hydroxyphenyl-1-ethanone, 1-2-fluoro-4-hydroxyphenyl ethan-1-one, 1-2-fluoro-4-hydroxy-phenyl ethanone, ethanone, 1-2-fluoro-4-hydroxyphenyl, 4'-hydroxy-2'-fluoroacetophenone
PubChem CID 2724912
IUPAC Name 1-(2-fluoro-4-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(O)C=C1F

Specifications

Melting Point 121°C to 123°C
Quantity 25 g
Beilstein 696642
Solubility Information Insoluble in water.
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 2'-Fluoro-4'-hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only