accessibility menu, dialog, popup

UserName

CAS RN 98541-64-1

H3CH3CCH3OONHOO

CAS RN 98541-64-1

IUPAC Name: tert-butyl N-(2-oxooxetan-3-yl)carbamate
Synonyms: tert-butyl N-(2-oxooxetan-3-yl)carbamate
Molecular Weight (g/mol): 187.2
Molecular Formula: C8H13NO4
InChi Key: HRJDEHQWXAPGBG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1COC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results