missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-serine beta-Lactone 98.0+%, TCI America™

Catalog No. B19955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19955G 5 g
B19951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19955G Supplier TCI America Supplier No. B19955G
Only null left

Chemical Identifiers

CAS 98541-64-1
Molecular Formula C8H13NO4
Molecular Weight (g/mol) 187.195
MDL Number MFCD01318414
InChI Key HRJDEHQWXAPGBG-YFKPBYRVSA-N
Synonym (S)-alpha-Boc-amino-beta-propiolactone, N-Boc-L-serine beta-Lactone, (S)-3-(tert-Butoxycarbonylamino)-2-oxetanone, (S)-alpha-tert-Butoxycarbonylamino-beta-propiolactone
PubChem CID 2763194
IUPAC Name tert-butyl N-[(3S)-2-oxooxetan-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1COC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 187.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-serine beta-Lactone
TSCA No
Recommended Storage Freezer