accessibility menu, dialog, popup

UserName

CAS RN 985-16-0

IUPAC Name: sodium (2S,5R,6R)-6-(2-ethoxynaphthalene-1-amido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: nafcillin sodium nafcillin sodium anhydrous
Molecular Weight (g/mol): 436.46
Molecular Formula: C21H21N2NaO5S
InChi Key: AYAPZOUDXCDGIF-FRFVDRIFSA-M
SMILES: [Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results