missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nafcillin sodium salt

Catalog No. AC461380050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461380050 5 g
AC461380010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461380050 Supplier Thermo Scientific Chemicals Supplier No. 461380050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 985-16-0 | C21H21N2NaO5S | 436.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 985-16-0
Molecular Formula C21H21N2NaO5S
Molecular Weight (g/mol) 436.46
MDL Number MFCD00056863
InChI Key AYAPZOUDXCDGIF-FRFVDRIFSA-M
Synonym 4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r
PubChem CID 131675018
IUPAC Name (2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O

Specifications

Melting Point >181.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 436.46
Chemical Name or Material Nafcillin sodium salt
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. In case of inadequate ventilation wear respiratory protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
EINECSNumber 213-574-4

RUO – Research Use Only