accessibility menu, dialog, popup

UserName

CAS RN 98-42-0

H3COH2NSOHOO

CAS RN 98-42-0

IUPAC Name: 3-amino-4-methoxybenzene-1-sulfonic acid
Synonyms: 3-amino-4-methoxybenzenesulfonic acid
Molecular Weight (g/mol): 203.21
Molecular Formula: C7H9NO4S
InChi Key: FLIOATBXVNLPLK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1N)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results