missing translation for 'onlineSavingsMsg'
Learn More

o-Anisidine-5-sulfonic Acid 95.0+%, TCI America™

Catalog No. A0491500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0491500G 500 g
A049125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0491500G Supplier TCI America Supplier No. A0491500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-42-0
Molecular Formula C7H9NO4S
Molecular Weight (g/mol) 203.212
MDL Number MFCD00035759
InChI Key FLIOATBXVNLPLK-UHFFFAOYSA-N
Synonym 4-methoxymetanilic acid, 2-anisidine-4-sulfonic acid, 2-methoxyaniline-5-sulfonic acid, benzenesulfonic acid, 3-amino-4-methoxy, o-anisidine-4-sulfonic acid, 2-methoxy-5-sulfoaniline, o-anisidine-p-sulfonic acid, 2-aminoanisole-4-sulfonic acid, benzenesulfonic acid, 3-amino-4-methoxyl, acmc-20ahhh
PubChem CID 66821
IUPAC Name 3-amino-4-methoxybenzenesulfonic acid
SMILES COC1=C(C=C(C=C1)S(=O)(=O)O)N

Specifications

Melting Point 305°C
Quantity 500 g
UN Number 2585
Formula Weight 203.21
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Anisidine-5-sulfonic Acid
EINECSNumber (3)-2037&(3)-2040
TSCA Yes