accessibility menu, dialog, popup

UserName

NH2HOSOHOO

CAS RN 98-37-3

IUPAC Name: 3-amino-4-hydroxybenzene-1-sulfonic acid
Synonyms: 2-aminophenol-4-sulfonic acid
Molecular Weight (g/mol): 189.19
Molecular Formula: C6H7NO4S
InChi Key: ULUIMLJNTCECJU-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results