missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzenesulfonic Acid Hydrate 98.0+%, TCI America™

Catalog No. A0399500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0399500G 500 g
A039925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0399500G Supplier TCI America Supplier No. A0399500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-37-3
Molecular Formula C6H7NO4S
Molecular Weight (g/mol) 189.185
MDL Number MFCD00007696
InChI Key ULUIMLJNTCECJU-UHFFFAOYSA-N
Synonym 2-Aminophenol-4-sulfonic Acid, 4-Hydroxymetanilic Acid
PubChem CID 7385
IUPAC Name 3-amino-4-hydroxybenzenesulfonic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)N)O

Specifications

Melting Point 220°C
Quantity 500 g
Formula Weight 189.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-hydroxybenzenesulfonic Acid Hydrate
EINECSNumber (3)-2029&(3)-3159
TSCA Yes